Tisk…

Anti-Succinic Acid Rat Polyclonal Antibody (Unconjugated)

Dodavatel: US Biological
S8008-01A
USBIS8008-01AEA 21020 CZK
USBIS8008-01A
Anti-Succinic Acid Rat Polyclonal Antibody (Unconjugated)
Protilátky
Type: Primary
Antigen: Succinic Acid
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity:
Objednat

Produktová specifikace

Název antigenu Succinic Acid (conjugated)
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita
Hostitel Rat
Izotyp IgG
ELISA Yes
Formulář lyophilized powder
Čištění purified by protein carrier (BSA) affinity column, and ammonium sulphate precipitation
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR