Tisk…

Anti-STRAP Rabbit Polyclonal Antibody

Dodavatel: US Biological
S7973-68
USBIS7973-68EA 19000 CZK
USBIS7973-68
Anti-STRAP Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: STRAP (serine/threonine kinase receptor associated protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR