Tisk…

Anti-Spexin Rabbit Polyclonal Antibody (Carboxyfluorescein)

Dodavatel: US Biological
S5401-91C
USBIS5401-91CEA 23810 CZK
USBIS5401-91C
Anti-Spexin Rabbit Polyclonal Antibody (Carboxyfluorescein)
Protilátky
Type: Primary
Antigen: Spexin
Clonality: Polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR