Tisk…

Anti-SOX8 Goat Polyclonal Antibody

Dodavatel: US Biological
S5364-25D
USBIS5364-25DEA 13470 CZK
USBIS5364-25D
Anti-SOX8 Goat Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: SOX8 (SRY (sex determining region Y)-box 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR