Tisk…

Anti-Salicylic Acid Sheep Polyclonal Antibody

Dodavatel: US Biological
S0058-19
USBIS0058-19EA 21700 CZK
USBIS0058-19
Anti-Salicylic Acid Sheep Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 15 ?g/ml

Type: Primary
Antigen: Salicylic Acid
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR