Tisk…

Anti-Rubella, E2 Mouse Monoclonal Antibody [clone: 9L637]

Dodavatel: US Biological
R9700-06C
USBIR9700-06CEA 19480 CZK
USBIR9700-06C
Anti-Rubella, E2 Mouse Monoclonal Antibody [clone: 9L637]
Protilátky
Type: Primary
Antigen: Rubella, E2
Clonality: monoclonal
Clone: 9L637
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Objednat

Produktová specifikace

Symbol antigenu Rubella, E2
Environmentally Preferable
Druh protilátky Primární
Klonovalita Monoclonal
Spojení Unconjugated
Klon 9L637
Reaktivita
Hostitel Mouse
Izotyp IgG1 kappa
Western blot Yes
Formát ascites
Formulář liquid
Skladovací pufr 0,02M Boric Acid, 5mM sodium Borate, 0,2M sodium chloride, pH 8.0+0,2, 0,1% sodium azide.
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR