Tisk…

Anti-Resistin-like Molecule, alpha Rabbit Polyclonal Antibody (Biotin)

Dodavatel: US Biological
R1587-07S
USBIR1587-07SEA 15120 CZK
USBIR1587-07S
Anti-Resistin-like Molecule, alpha Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:5000-1:10000); Western Blot: 1:5000-1:10000

Type: Primary
Antigen: Resistin-like Molecule, alpha
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR