Tisk…

Anti-PPM1F Rabbit Polyclonal Antibody

Dodavatel: US Biological
P9107-19T
USBIP9107-19TEA 19000 CZK
USBIP9107-19T
Anti-PPM1F Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: PPM1F (protein phosphatase, Mg2+/Mn2+ dependent, 1F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR