Tisk…

Anti-PPM1E Rabbit Polyclonal Antibody (Biotin)

Dodavatel: US Biological
P9107-19H-BIOT
USBIP9107-19H-BIOTEA 25170 CZK
USBIP9107-19H-BIOT
Anti-PPM1E Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: PPM1E (protein phosphatase, Mg2+/Mn2+ dependent, 1E)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR