Tisk…

Anti-Protein Gene Product 9.5 Rabbit Polyclonal Antibody

Dodavatel: US Biological
P9102-70G
USBIP9102-70GEA 12300 CZK
USBIP9102-70G
Anti-Protein Gene Product 9.5 Rabbit Polyclonal Antibody
Protilátky
  • Recommended dilution: Immunochemistry: 1:500-1:1000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Protein Gene Product 9.5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat
Objednat

Produktová specifikace

Název antigenu Protein Gene Product 9.5
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Pig,Human,Rat,Mouse
Hostitel Rabbit
Izotyp IgG
Western blot Yes
Formát Serum
Formulář liquid
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR