Tisk…

Anti-Prostate Specific Gene-1 Rabbit Polyclonal Antibody

Dodavatel: US Biological
P9054-77B
USBIP9054-77BEA 16520 CZK
USBIP9054-77B
Anti-Prostate Specific Gene-1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Prostate Specific Gene-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR