Tisk…

Anti-Pro-Platelet Basic Protein Chicken Polyclonal Antibody

Dodavatel: US Biological
P9052-07
USBIP9052-07EA 20760 CZK
USBIP9052-07
Anti-Pro-Platelet Basic Protein Chicken Polyclonal Antibody
Protilátky
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Pro-Platelet Basic Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR