Tisk…

Anti-Phosphorylase Kinase Subunit gamma 1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: US Biological
P4075-85-AP
USBIP4075-85-APEA 25170 CZK
USBIP4075-85-AP
Anti-Phosphorylase Kinase Subunit gamma 1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: Phosphorylase Kinase Subunit gamma 1
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR