Tisk…

Anti-Pericentriolar Material 1 Protein Rabbit Polyclonal Antibody

Dodavatel: US Biological
P3340-53B
USBIP3340-53BEA 19000 CZK
USBIP3340-53B
Anti-Pericentriolar Material 1 Protein Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: Pericentriolar Material 1 Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR