Tisk…

Anti-OXT Goat Polyclonal Antibody

Dodavatel: US Biological
O8250-05A
USBIO8250-05AEA 18230 CZK
USBIO8250-05A
Anti-OXT Goat Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: OXT (oxytocin/neurophysin I prepropeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR