Tisk…

Anti-OPTN Rabbit Polyclonal Antibody

Dodavatel: US Biological
O7037-02B
USBIO7037-02BEA 18000 CZK
USBIO7037-02B
Anti-OPTN Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: OPTN (optineurin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR