Tisk…

Anti-NUB1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Dodavatel: US Biological
N8574-10-FITC
USBIN8574-10-FITCEA 25170 CZK
USBIN8574-10-FITC
Anti-NUB1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: NUB1 (negative regulator of ubiquitin-like proteins 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR