Tisk…

Anti-N-myc Downstream Regulated Gene 1 Protein Rabbit Polyclonal Antibody (Biotin)

Dodavatel: US Biological
N2915-52N-BIOT
USBIN2915-52N-BIOTEA 25170 CZK
USBIN2915-52N-BIOT
Anti-N-myc Downstream Regulated Gene 1 Protein Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: N-myc Downstream Regulated Gene 1 Protein
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR