Tisk…

Anti-MRII Rabbit Polyclonal Antibody

Dodavatel: US Biological
M3955-30C
USBIM3955-30CEA 19000 CZK
USBIM3955-30C
Anti-MRII Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: MRII
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR