Tisk…

Anti-METTL7B Rabbit Polyclonal Antibody

Dodavatel: US Biological
M3599-02A
USBIM3599-02AEA 21440 CZK
USBIM3599-02A
Anti-METTL7B Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Western Blot: 2-4?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: METTL7B (methyltransferase like 7B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR