Tisk…

Anti-Methionine-tRNA Synthetase, Mitochondrial Rabbit Polyclonal Antibody

Dodavatel: US Biological
M3022-56C
USBIM3022-56CEA 19000 CZK
USBIM3022-56C
Anti-Methionine-tRNA Synthetase, Mitochondrial Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: Methionine-tRNA Synthetase, Mitochondrial
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR