Tisk…

Anti-Matrix Metalloproteinase 9 Rabbit Polyclonal Antibody

Dodavatel: US Biological
M2425-01N
USBIM2425-01NEA 20280 CZK
USBIM2425-01N
Anti-Matrix Metalloproteinase 9 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Matrix Metalloproteinase 9
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR