Tisk…

Anti-L-Lactate Dehydrogenase, LDH-1 Sheep Polyclonal Antibody

Dodavatel: US Biological
L1012-70A
USBIL1012-70AEA 23290 CZK
USBIL1012-70A
Anti-L-Lactate Dehydrogenase, LDH-1 Sheep Polyclonal Antibody
Protilátky
Type: Primary
Antigen: L-Lactate Dehydrogenase, LDH-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR