Tisk…

Anti-KDM2B Rabbit Polyclonal Antibody (FITC (Fluorescein))

Dodavatel: US Biological
K0138-40B-FITC
USBIK0138-40B-FITCEA 25170 CZK
USBIK0138-40B-FITC
Anti-KDM2B Rabbit Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: KDM2B (lysine (K)-specific demethylase 2B)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR