Tisk…

Anti-Interleukin-1 Mouse Monoclonal Antibody [clone: 9L377]

Dodavatel: US Biological
I7663-25G3
USBII7663-25G3EA 21590 CZK
USBII7663-25G3
Anti-Interleukin-1 Mouse Monoclonal Antibody [clone: 9L377]
Protilátky
Recommended dilution: ELISA: 5 ?g/ml; Immunochemistry: 10-100 ?g/mlRadio Immunoassay: 20-30ng/ml

Type: Primary
Antigen: Interleukin-1
Clonality: monoclonal
Clone: 9L377
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR