Tisk…

Anti-IRS1 Mouse Monoclonal Antibody [clone: 10g79]

Dodavatel: US Biological
I7661-27F
USBII7661-27FEA 13960 CZK
USBII7661-27F
Anti-IRS1 Mouse Monoclonal Antibody [clone: 10g79]
Protilátky
Recommended dilution: ELISA: 1-2?g/ml

Type: Primary
Antigen: IRS1 (insulin receptor substrate 1)
Clonality: monoclonal
Clone: 10g79
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR