Tisk…

Anti-Histone H3, monomethyl Rabbit polyclonal antibody

Dodavatel: US Biological
H5110-11G2
USBIH5110-11G2EA 18850 CZK
USBIH5110-11G2
Anti-Histone H3, monomethyl Rabbit polyclonal antibody
Protilátky
Recommended dilution: Immunoblot: A 1:1000

Type: Primary
Antigen: Histone H3, monomethyl
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR