Tisk…

Anti-GRO beta Rabbit Polyclonal Antibody

Dodavatel: US Biological
G8975-22C
USBIG8975-22CEA 22070 CZK
USBIG8975-22C
Anti-GRO beta Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 0.5 ?g/ml; Western Blot: 0.1-0.2?g/ml; Functional Assays: 0.5-1 ?g/ml

Type: Primary
Antigen: GRO beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR