Tisk…

Anti-Glutathione S Transferase, pi Sheep polyclonal antibody

Dodavatel: US Biological
G8135-26E
USBIG8135-26EEA 19620 CZK
USBIG8135-26E
Anti-Glutathione S Transferase, pi Sheep polyclonal antibody
Protilátky
Recommended dilution: Western Blot: 1:100-1:400

Type: Tag
Antigen: Glutathione S Transferase, pi
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR