Tisk…

Anti-EPM2A Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Dodavatel: US Biological
E3384-75G-PE
USBIE3384-75G-PEEA 25170 CZK
USBIE3384-75G-PE
Anti-EPM2A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Protilátky
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: EPM2A (epilepsy, progressive myoclonus type 2A, Lafora disease (laforin))
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu EPM2A
Environmentally Preferable
Název antigenu Epilepsy progressive myoclonus type 2A Lafora disease (laforin)
Druh protilátky Primární
Klonovalita Polyclonal
Spojení PE (Phycoerythrin)
Reaktivita Human
Hostitel Rabbit
Izotyp IgG
ELISA Yes
Epitop N-terminal
Formulář liquid
Čištění purified by Protein A chromatography
Skladovací pufr PBS, pH 7,2
Skladovací teplota Cool
Transportní teplota Ice
Testované aplikace Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR