Tisk…

Anti-Encephalitis, St. Louis Mouse monoclonal antibody [clone: 10D421]

Dodavatel: US Biological
E2280-50A
USBIE2280-50AEA 22070 CZK
USBIE2280-50A
Anti-Encephalitis, St. Louis Mouse monoclonal antibody [clone: 10D421]
Protilátky
Type: Primary
Antigen: Encephalitis, St. Louis
Clonality: monoclonal
Clone: 10D421
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR