Tisk…

Anti-DOK2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Dodavatel: US Biological
D8080-02H-FITC
USBID8080-02H-FITCEA 25170 CZK
USBID8080-02H-FITC
Anti-DOK2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: DOK2 (docking protein 2, 56kDa)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR