Tisk…

Anti-Delta-like 1 Mouse monoclonal antibody [clone: 7H56]

Dodavatel: US Biological
D2727-75
USBID2727-75EA 23610 CZK
USBID2727-75
Anti-Delta-like 1 Mouse monoclonal antibody [clone: 7H56]
Protilátky
Recommended dilution: Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: Delta-like 1
Clonality: monoclonal
Clone: 7H56
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR