Tisk…

Anti-CCNE1 Rabbit Polyclonal Antibody (Biotin)

Dodavatel: US Biological
C8455-01K-BIOT
USBIC8455-01K-BIOTEA 25170 CZK
USBIC8455-01K-BIOT
Anti-CCNE1 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: CCNE1 (cyclin E1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR