Tisk…

Anti-CDC25A Rabbit Polyclonal Antibody

Dodavatel: US Biological
C2560-01J
USBIC2560-01JEA 14550 CZK
USBIC2560-01J
Anti-CDC25A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR