Tisk…

Anti-SELE Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 10K40]

Dodavatel: US Biological
C2414-93A
USBIC2414-93AEA 18630 CZK
USBIC2414-93A
Anti-SELE Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 10K40]
Protilátky
Do not freeze!

Type: Primary
Antigen: SELE (selectin E)
Clonality: Monoclonal
Clone: 10K40
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR