Tisk…

Anti-CD59 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-43]

Dodavatel: US Biological
C2413-22F
USBIC2413-22FEA 17290 CZK
USBIC2413-22F
Anti-CD59 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-43]
Protilátky
Type: Primary
Antigen: CD59 (CD59 molecule, complement regulatory protein)
Clonality: Monoclonal
Clone: MEM-43
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu CD59
Environmentally Preferable
Název antigenu CD59 molecule, complement regulatory Protein
Druh protilátky Primární
Klonovalita Monoclonal
Spojení FITC (Fluorescein Isothiocyanate)
Klon MEM-43
Reaktivita Human
Hostitel Mouse
Izotyp IgG2a
Průtoková cytometrie Yes
Formulář liquid
Čištění purified by Protein A chromatography
Skladovací pufr PBS, pH 7,4, BSA, 15mM sodium azide.
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR