Tisk…

Anti-Catenin, beta Mouse Monoclonal Antibody [clone: 6D207]

Dodavatel: US Biological
C2069-52E
USBIC2069-52EEA 19220 CZK
USBIC2069-52E
Anti-Catenin, beta Mouse Monoclonal Antibody [clone: 6D207]
Protilátky
Type: Primary
Antigen: Catenin, beta
Clonality: Monoclonal
Clone: 6D207
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat,
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR