Tisk…

Anti-CPT2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Dodavatel: US Biological
C1385-50B-FITC
USBIC1385-50B-FITCEA 25170 CZK
USBIC1385-50B-FITC
Anti-CPT2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: CPT2 (carnitine palmitoyltransferase 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR