Tisk…

Anti-Calcitonin Gene Related Peptide Rabbit polyclonal antibody unconjugated

Dodavatel: US Biological
C0115-02K
USBIC0115-02KEA 22640 CZK
USBIC0115-02K
Anti-Calcitonin Gene Related Peptide Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: Calcitonin Gene Related Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR