Tisk…

Anti-Bu-1 Mouse Monoclonal Antibody (Biotin) [clone: 5K98]

Dodavatel: US Biological
B2788-45B
USBIB2788-45BEA 15550 CZK
USBIB2788-45B
Anti-Bu-1 Mouse Monoclonal Antibody (Biotin) [clone: 5K98]
Protilátky
Type: Primary
Antigen: Bu-1
Clonality: monoclonal
Clone: 5K98
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR