Tisk…

Anti-BAI3 Mouse monoclonal antibody unconjugated [clone: 8H174]

Dodavatel: US Biological
B2704-12C
USBIB2704-12CEA 22440 CZK
USBIB2704-12C
Anti-BAI3 Mouse monoclonal antibody unconjugated [clone: 8H174]
Protilátky
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Immunochemistry: 25 ?g/ml

Type: Primary
Antigen: BAI3 (brain-specific angiogenesis inhibitor 3)
Clonality: monoclonal
Clone: 8H174
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR