Tisk…

Anti-BRAF Rabbit Polyclonal Antibody (Biotin)

Dodavatel: US Biological
B2680-01J-BIOT
USBIB2680-01J-BIOTEA 25170 CZK
USBIB2680-01J-BIOT
Anti-BRAF Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR