Tisk…

Anti-BMPR1A Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Dodavatel: US Biological
B2553-62-APC
USBIB2553-62-APCEA 25170 CZK
USBIB2553-62-APC
Anti-BMPR1A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: BMPR1A (bone morphogenetic protein receptor, type IA)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR