Tisk…

Anti-BCL2L2 Mouse monoclonal antibody unconjugated [clone: 12K13]

Dodavatel: US Biological
B0003-61B
USBIB0003-61BEA 22810 CZK
USBIB0003-61B
Anti-BCL2L2 Mouse monoclonal antibody unconjugated [clone: 12K13]
Protilátky
Type: Primary
Antigen: BCL2L2 (BCL2-like 2)
Clonality: monoclonal
Clone: 12K13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR