Tisk…

Anti-B. anthracis SA Mouse Monoclonal Antibody [clone: 3G302A]

Dodavatel: US Biological
B0003-05G3
USBIB0003-05G3EA 32460 CZK
USBIB0003-05G3
Anti-B. anthracis SA Mouse Monoclonal Antibody [clone: 3G302A]
Protilátky
Type: Primary
Antigen: B. anthracis SA (Bacillus anthracis spore antigen)
Clonality: monoclonal
Clone: 3G302A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR