Tisk…

Anti-B Rabbit Polyclonal Antibody

Dodavatel: US Biological
B0002-15
USBIB0002-15EA 10620 CZK
USBIB0002-15
Anti-B Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.1-1.0 ?g/ml

Type: Primary
Antigen: B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR