Tisk…

Anti-Arginine, symmetric, dimethyl Rabbit Polyclonal Antibody

Dodavatel: US Biological
A3550-07B
USBIA3550-07BEA 26480 CZK
USBIA3550-07B
Anti-Arginine, symmetric, dimethyl Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: Arginine, symmetric, dimethyl
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Produktová specifikace

Symbol antigenu Arg
Název antigenu Arginine
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Mouse
Hostitel Rabbit
Izotyp IgG
Western blot Yes
Modifikace dimethyl-ASYM24
Formát Serum
Formulář liquid
Skladovací pufr PBS, 0,05% sodium azide, 30% glycerol.
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR