Tisk…
Cell Culture Media, Ames' Media
Dodavatel: US Biological
A1372-25A-100L
USBIA1372-25A-100LEA 24010 CZK
USBIA1372-25A-100L
Cell Culture Media, Ames' Media
Média kultivační
Ames' medium was formulated to support retinal tissue in relatively short-term culture.
  • White to pale yellow powder
  • Homogeneous, free flowing powder
Rabbit retina has been incubated in Ames' medium for over 2 days with its metabolism and electrical responses to light stimuli well maintained. This mixture is a medium of choice for maintaining central nervous system tissue in vitro.