Tisk…

Anti-AKT1/2 Rabbit Polyclonal Antibody

Dodavatel: US Biological
A1125-23A
USBIA1125-23AEA 15350 CZK
USBIA1125-23A
Anti-AKT1/2 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 2?g/ml

Type: Primary
Antigen: AKT1/2 (v-akt murine thymoma viral oncogene homolog 1/2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR